accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OIO

CAS RN 306934-90-7

IUPAC Name: 7-iodo-3,4-dihydro-2H-1,5-benzodioxepine
Synonyms: 7-iodo-3,4-dihydro-2H-1,5-benzodioxepine
Molecular Weight (g/mol): 276.07
Molecular Formula: C9H9IO2
InChi Key: CIZHAPZOOAJDCN-UHFFFAOYSA-N
SMILES: IC1=CC2=C(OCCCO2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results