accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3SiOH3COH3COH3C

CAS RN 3069-19-0

IUPAC Name: hexyltrimethoxysilane
Synonyms: hexyltrimethoxysilane
Molecular Weight (g/mol): 206.36
Molecular Formula: C9H22O3Si
InChi Key: CZWLNMOIEMTDJY-UHFFFAOYSA-N
SMILES: CCCCCC[Si](OC)(OC)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results