accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COSiNHOH3COCH3

CAS RN 3068-76-6

IUPAC Name: N-[3-(trimethoxysilyl)propyl]aniline
Synonyms: N-[3-(trimethoxysilyl)propyl]aniline
Molecular Weight (g/mol): 255.39
Molecular Formula: C12H21NO3Si
InChi Key: KBJFYLLAMSZSOG-UHFFFAOYSA-N
SMILES: CO[Si](CCCNC1=CC=CC=C1)(OC)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results