accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COOCH3

CAS RN 30673-60-0

IUPAC Name: propyl decanoate
Synonyms: propyl decanoate
Molecular Weight (g/mol): 214.35
Molecular Formula: C13H26O2
InChi Key: OVFMRFMJVFDSAA-UHFFFAOYSA-N
SMILES: CCCCCCCCCC(=O)OCCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results