accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 30650-90-9

H3COOH3CNOO

CAS RN 30650-90-9

IUPAC Name: ethyl 3-methyl-4-nitrobenzoate
Synonyms: ethyl 3-methyl-4-nitrobenzoate
Molecular Weight (g/mol): 209.2
Molecular Formula: C10H11NO4
InChi Key: OAXJZQMFWBIRKF-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(C(C)=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results