accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

SSHNN

CAS RN 306281-11-8

IUPAC Name: 7-thia-9,11-diazatricyclo[6.4.0.0²,⁶]dodeca-1(8),2(6),9-triene-12-thione
Synonyms: 7-thia-9,11-diazatricyclo[6.4.0.0²,⁶]dodeca-1(8),2(6),9-triene-12-thione
Molecular Weight (g/mol): 208.3
Molecular Formula: C9H8N2S2
InChi Key: UZIFVIYMKOHMQD-UHFFFAOYSA-N
SMILES: S=C1NC=NC2=C1C1=C(CCC1)S2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results