accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3054-95-3

H3COOCH3CH2

CAS RN 3054-95-3

IUPAC Name: 3,3-diethoxyprop-1-ene
Synonyms: 1-propene, 3,3-diethoxy-
Molecular Weight (g/mol): 130.19
Molecular Formula: C7H14O2
InChi Key: MCIPQLOKVXSHTD-UHFFFAOYSA-N
SMILES: CCOC(OCC)C=C
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)