accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3047-32-3

IUPAC Name: (3-ethyloxetan-3-yl)methanol
Synonyms: (3-ethyloxetan-3-yl)methanol
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: UNMJLQGKEDTEKJ-UHFFFAOYSA-N
SMILES: CCC1(CO)COC1
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)