accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

SNNHOCl

CAS RN 304685-87-8

IUPAC Name: 2-chloro-N-{3-cyano-4H,5H,6H-cyclopenta[b]thiophen-2-yl}acetamide
Synonyms: 2-chloro-N-{3-cyano-4H,5H,6H-cyclopenta[b]thiophen-2-yl}acetamide
Molecular Weight (g/mol): 240.71
Molecular Formula: C10H9ClN2OS
InChi Key: IFPYPHZLEFBRSS-UHFFFAOYSA-N
SMILES: ClCC(=O)NC1=C(C#N)C2=C(CCC2)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results