accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CNNCH3FPFFFFF

CAS RN 304680-35-1

IUPAC Name: 1-hexyl-3-methyl-1H-imidazol-3-ium; hexafluoro-λ⁵-phosphanuide
Synonyms: 1-hexyl-3-methylimidazolium hexafluorophosphate
Molecular Weight (g/mol): 312.24
Molecular Formula: C10H19F6N2P
InChi Key: YPWSSSRXUOQNMQ-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.CCCCCCN1C=C[N+](C)=C1
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results