accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 304669-35-0

CH3SiH3COOH

CAS RN 304669-35-0

IUPAC Name: (3,4-dihydro-2H-pyran-6-yl)dimethylsilanol
Synonyms: 5,6-dihydro-4H-pyran-2-yldimethylsilanol
Molecular Weight (g/mol): 158.27
Molecular Formula: C7H14O2Si
InChi Key: ZMDNGTLKIVNNCF-UHFFFAOYSA-N
SMILES: C[Si](C)(O)C1=CCCCO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results