accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FFFNNH

CAS RN 30459-17-7

IUPAC Name: 1-[4-(trifluoromethyl)phenyl]piperazine
Synonyms: 1-[4-(trifluoromethyl)phenyl]piperazine
Molecular Weight (g/mol): 230.23
Molecular Formula: C11H13F3N2
InChi Key: IBQMAPSJLHRQPE-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(C=C1)N1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results