accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 30450-62-5

IUPAC Name: 7-nitro-1,2,3,4-tetrahydroquinoline
Synonyms: 7-nitro-1,2,3,4-tetrahydroquinoline
Molecular Weight (g/mol): 178.19
Molecular Formula: C9H10N2O2
InChi Key: WSWMGHRLUYADNA-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC2=C(CCCN2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results