accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3034-62-6

IUPAC Name: 2-iodo-1H-imidazole
Synonyms: 2-iodo-1H-imidazole
Molecular Weight (g/mol): 193.98
Molecular Formula: C3H3IN2
InChi Key: MHHDMDLNVVCTAJ-UHFFFAOYSA-N
SMILES: IC1=NC=CN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results