accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ONOSBrN

CAS RN 3034-48-8

IUPAC Name: 2-bromo-5-nitro-1,3-thiazole
Synonyms: 2-bromo-5-nitro-1,3-thiazole
Molecular Weight (g/mol): 209.02
Molecular Formula: C3HBrN2O2S
InChi Key: ANIJFZVZXZQFDH-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CN=C(Br)S1
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results