accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3033-82-7

H3CClN

CAS RN 3033-82-7

IUPAC Name: 8-chloro-2-methylquinoline
Synonyms: 8-chloro-2-methylquinoline
Molecular Weight (g/mol): 177.63
Molecular Formula: C10H8ClN
InChi Key: VVLYDFPOGMTMFJ-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=CC=C(Cl)C2=N1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)