accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

HNNOOHOHOH2O

CAS RN 303180-11-2

IUPAC Name: 1H-pyrazole-3,5-dicarboxylic acid hydrate
Synonyms: 1H-pyrazole-3,5-dicarboxylic acid hydrate
Molecular Weight (g/mol): 174.11
Molecular Formula: C5H6N2O5
InChi Key: GLINCONFUZIMCN-UHFFFAOYSA-N
SMILES: O.OC(=O)C1=CC(=NN1)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results