accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3025-95-4

H3CONHOHO

CAS RN 3025-95-4

IUPAC Name: 3-acetamidopropanoic acid
Synonyms: N-acetyl-β-alanine
Molecular Weight (g/mol): 131.13
Molecular Formula: C5H9NO3
InChi Key: LJLLAWRMBZNPMO-UHFFFAOYSA-N
SMILES: CC(=O)NCCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)