accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 301652-23-3

CH3SnH3CCH3NH3C

CAS RN 301652-23-3

IUPAC Name: 4-methyl-2-(tributylstannyl)pyridine
Synonyms: 4-methyl-2-(tributylstannyl)pyridine
Molecular Weight (g/mol): 382.18
Molecular Formula: C18H33NSn
InChi Key: SHMPGQWVDZTZBX-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)C1=NC=CC(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results