accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3010-03-5

IUPAC Name: 2-(piperidin-1-yl)acetonitrile
Synonyms: 2-(piperidin-1-yl)acetonitrile
Molecular Weight (g/mol): 124.19
Molecular Formula: C7H12N2
InChi Key: CLVBVRODHJFTGF-UHFFFAOYSA-N
SMILES: N#CCN1CCCCC1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results