accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3001-15-8

IUPAC Name: 4,4'-diiodo-1,1'-biphenyl
Synonyms: 4,4'-diiodo-1,1'-biphenyl
Molecular Weight (g/mol): 406.01
Molecular Formula: C12H8I2
InChi Key: GPYDMVZCPRONLW-UHFFFAOYSA-N
SMILES: IC1=CC=C(C=C1)C1=CC=C(I)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results