accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 299919-85-0

CH3SONSHN

CAS RN 299919-85-0

IUPAC Name: 11-methyl-4-phenyl-5-sulfanylidene-8-thia-4,6-diazatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7)-dien-3-one
Synonyms: 11-methyl-4-phenyl-5-sulfanylidene-8-thia-4,6-diazatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7)-dien-3-one
Molecular Weight (g/mol): 328.45
Molecular Formula: C17H16N2OS2
InChi Key: IVSIHDBMSUYSAH-UHFFFAOYNA-N
SMILES: CC1CCC2=C(C1)SC1=C2C(=O)N(C(=S)N1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results