accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3ONHNNH2

CAS RN 29958-14-3

IUPAC Name: N-(5-aminopyridin-2-yl)acetamide
Synonyms: N-(5-aminopyridin-2-yl)acetamide
Molecular Weight (g/mol): 151.17
Molecular Formula: C7H9N3O
InChi Key: SXGURHSLXPDJBC-UHFFFAOYSA-N
SMILES: CC(=O)NC1=NC=C(N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results