accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ClICl

CAS RN 29898-32-6

IUPAC Name: 2,4-dichloro-1-iodobenzene
Synonyms: 2,4-dichloro-1-iodobenzene
Molecular Weight (g/mol): 272.89
Molecular Formula: C6H3Cl2I
InChi Key: ZQKJCBDCOGLKCQ-UHFFFAOYSA-N
SMILES: ClC1=CC=C(I)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results