accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 29767-20-2

CH3OOHOCH3OOOOOOOOSOHHO

CAS RN 29767-20-2

IUPAC Name: 16-{[7,8-dihydroxy-2-(thiophen-2-yl)-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-6-yl]oxy}-10-(4-hydroxy-3,5-dimethoxyphenyl)-4,6,13-trioxatetracyclo[7.7.0.0³,⁷.0¹¹,¹⁵]hexadeca-1,3(7),8-trien-12-one
Synonyms: teniposide
Molecular Weight (g/mol): 656.66
Molecular Formula: C32H32O13S
InChi Key: NRUKOCRGYNPUPR-UHFFFAOYNA-N
SMILES: COC1=CC(=CC(OC)=C1O)C1C2C(COC2=O)C(OC2OC3COC(OC3C(O)C2O)C2=CC=CS2)C2=CC3=C(OCO3)C=C12
Molecular Weight (g/mol) 
  • (4)
  • (2)
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (1)
  • (4)
  • (1)
  • (4)
  • (2)
  • (4)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)