accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 29681-57-0

CH3H3CCH3SiCH3H3C

CAS RN 29681-57-0

IUPAC Name: tert-butyldimethylsilyl
Synonyms: tert-butyldimethylsilyl group
Molecular Weight (g/mol): 115.27
Molecular Formula: C6H15Si
InChi Key: ILMRJRBKQSSXGY-UHFFFAOYSA-N
SMILES: C[Si](C)C(C)(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results