accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 29668-44-8

IUPAC Name: 2,3-dihydro-1,4-benzodioxine-6-carbaldehyde
Synonyms: 2,3-dihydro-1,4-benzodioxine-6-carbaldehyde
Molecular Weight (g/mol): 164.16
Molecular Formula: C9H8O3
InChi Key: CWKXDPPQCVWXAG-UHFFFAOYSA-N
SMILES: O=CC1=CC=C2OCCOC2=C1
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (3)
  • (1)
  • (3)
Boiling Point 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)