accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

AgOOFFF

CAS RN 2966-50-9

IUPAC Name: silver(1+) trifluoroacetate
Synonyms: silver(1+) trifluoroacetate
Molecular Weight (g/mol): 220.88
Molecular Formula: C2AgF3O2
InChi Key: KZJPVUDYAMEDRM-UHFFFAOYSA-M
SMILES: [Ag+].[O-]C(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results