accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 2965-22-2

IUPAC Name: methyl 2-fluoro-5-nitrobenzoate
Synonyms: methyl 2-fluoro-5-nitrobenzoate
Molecular Weight (g/mol): 199.14
Molecular Formula: C8H6FNO4
InChi Key: JZLONOOYIXEAHM-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(F)C=CC(=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results