accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 29558-77-8

IUPAC Name: 4'-bromo-[1,1'-biphenyl]-4-ol
Synonyms: 4'-bromo-[1,1'-biphenyl]-4-ol
Molecular Weight (g/mol): 249.11
Molecular Formula: C12H9BrO
InChi Key: ARUBXNBYMCVENE-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C1=CC=C(Br)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results