accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3H3CSnCH3CH3CH3H3CCH3H3C

CAS RN 2949-42-0

IUPAC Name: tetrakis(propan-2-yl)stannane
Synonyms: stannane, tetraisopropyl-
Molecular Weight (g/mol): 291.07
Molecular Formula: C12H28Sn
InChi Key: NJASUIDIZMMYED-UHFFFAOYSA-N
SMILES: CC(C)[Sn](C(C)C)(C(C)C)C(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)