accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 2946-17-0

IUPAC Name: 5-methoxy-2-methyl-1,3-benzoselenazole
Synonyms: 5-methoxy-2-methyl-1,3-benzoselenazole
Molecular Weight (g/mol): 226.15
Molecular Formula: C9H9NOSe
InChi Key: LVIYTMFDCXRSDL-UHFFFAOYSA-N
SMILES: COC1=CC=C2[Se]C(C)=NC2=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results