accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ONOSN

CAS RN 2942-07-6

IUPAC Name: 5-nitro-1,3-benzothiazole
Synonyms: 5-nitro-1,3-benzothiazole
Molecular Weight (g/mol): 180.18
Molecular Formula: C7H4N2O2S
InChi Key: AEUQLELVLDMMKB-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2SC=NC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results