accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COOSH

CAS RN 2935-90-2

IUPAC Name: methyl 3-sulfanylpropanoate
Synonyms: methyl 3-mercaptopropionate
Molecular Weight (g/mol): 120.17
Molecular Formula: C4H8O2S
InChi Key: LDTLDBDUBGAEDT-UHFFFAOYSA-N
SMILES: COC(=O)CCS
Molecular Weight (g/mol) 
  • (2)
  • (6)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (6)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)