accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 2923-17-3

LiOOFFF

CAS RN 2923-17-3

IUPAC Name: lithium(1+) trifluoroacetate
Synonyms: lithium(1+) trifluoroacetate
Molecular Weight (g/mol): 119.96
Molecular Formula: C2F3LiO2
InChi Key: HSFDLPWPRRSVSM-UHFFFAOYSA-M
SMILES: [Li+].[O-]C(=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results