accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FFFSOONSOOFFFCu

CAS RN 291300-50-0

IUPAC Name: λ¹-copper(1+) trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Synonyms: λ¹-copper(1+) bistriflylimide anion
Molecular Weight (g/mol): 343.68
Molecular Formula: C2CuF6NO4S2
InChi Key: MSFHCZUIBARZOS-UHFFFAOYSA-N
SMILES: [Cu+].FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)