accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3H3COH3CH3CNO

CAS RN 2896-70-0

IUPAC Name: (2,2,6,6-tetramethyl-4-oxopiperidin-1-yl)oxidanyl
Synonyms: 2,2,6,6-tetramethyl-4-oxopiperidin-1-yloxidanyl
Molecular Weight (g/mol): 170.23
Molecular Formula: C9H16NO2
InChi Key: WSGDRFHJFJRSFY-UHFFFAOYSA-N
SMILES: CC1(C)CC(=O)CC(C)(C)N1[O]
Quantity 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)