CAS RN 2895-21-8
CAS RN 2895-21-8
IUPAC Name:
2-chloro-N-(propan-2-yl)acetamide
Synonyms:
2-chloro-N-isopropylacetamide
Molecular Weight (g/mol):
135.59
Molecular Formula:
C5H10ClNO
InChi Key:
GYPNJSBBOATUPK-UHFFFAOYSA-N
SMILES:
CC(C)NC(=O)CCl
1
–
1
of
1
results
1
N-Isopropyl-2-chloroacetamide, 97%, Thermo Scientific™
CAS: 2895-21-8 Molecular Formula: C5H10ClNO Molecular Weight (g/mol): 135.591 MDL Number: MFCD00068034 InChI Key: GYPNJSBBOATUPK-UHFFFAOYSA-N Synonym: 2-chloro-n-isopropylacetamide,n-isopropyl-2-chloroacetamide,2-chloro-n-1-methylethyl acetamide,n-isopropyl chloroacetamide,2-chloro-n-propan-2-yl acetamide,acetamide, 2-chloro-n-1-methylethyl,n-chloroacetyl isopropylamine,2-chloro-n-methylethyl acetamide,acmc-1cpp2,2-chloro-n-isopropyl acetamide PubChem CID: 76171 IUPAC Name: 2-chloro-N-propan-2-ylacetamide SMILES: CC(C)NC(=O)CCl