accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 289039-84-5

CH3OOH2NICl

CAS RN 289039-84-5

IUPAC Name: methyl 2-amino-5-chloro-3-iodobenzoate
Synonyms: methyl 2-amino-5-chloro-3-iodobenzoate
Molecular Weight (g/mol): 311.5
Molecular Formula: C8H7ClINO2
InChi Key: VTDNWMZYPVQPCN-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Cl)=CC(I)=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results