accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 289039-53-8

H3CBrBr

CAS RN 289039-53-8

IUPAC Name: 1,4-dibromo-2-ethylbenzene
Synonyms: 1,4-dibromo-2-ethylbenzene
Molecular Weight (g/mol): 263.96
Molecular Formula: C8H8Br2
InChi Key: OMHJVVBCCMLFMY-UHFFFAOYSA-N
SMILES: CCC1=C(Br)C=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results