accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OHOOO

CAS RN 2861-28-1

IUPAC Name: 2-(2H-1,3-benzodioxol-5-yl)acetic acid
Synonyms: 2H-1,3-benzodioxol-5-ylacetic acid
Molecular Weight (g/mol): 180.16
Molecular Formula: C9H8O4
InChi Key: ODVLMCWNGKLROU-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=C2OCOC2=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (3)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)