accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 28588-74-1

CH3OHS

CAS RN 28588-74-1

IUPAC Name: 2-methylfuran-3-thiol
Synonyms: 2-methyl-3-furanthiol
Molecular Weight (g/mol): 114.16
Molecular Formula: C5H6OS
InChi Key: RUYNUXHHUVUINQ-UHFFFAOYSA-N
SMILES: CC1=C(S)C=CO1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results