accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 28529-99-9

NaNaBrIrBrBrBrBrBr

CAS RN 28529-99-9

IUPAC Name: disodium hexabromoiridiumdiuide
Synonyms: disodium hexabromoiridiumdiuide
Molecular Weight (g/mol): 717.62
Molecular Formula: Br6IrNa2
InChi Key: XUAIOVIZUXDJGC-UHFFFAOYSA-H
SMILES: [Na+].[Na+].Br[Ir--](Br)(Br)(Br)(Br)Br
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results