CAS RN 285132-91-4
CAS RN 285132-91-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
4-Chlorophenol-d4, TRC
CAS: 285132-91-4 Molecular Formula: C6 2H4 H Cl O Molecular Weight (g/mol): 132.58 Synonym: Phen-2,3,5,6-d4-ol, 4-chloro- (9CI),4-Chlorophenol-D4,p-Chlorophenol-d4,4-Chlorophenol D4 (phenyl D4) IUPAC Name: 4-chloro-2,3,5,6-tetradeuteriophenol SMILES: [2H]c1c([2H])c(Cl)c([2H])c([2H])c1O
2
4-Chlorophenol-2,3,5,6-d4, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.