accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 28503-70-0

TiCH3OCH3OCH3OCH3O

CAS RN 28503-70-0

IUPAC Name: titanium(4+) tetrakis(2-methylbenzen-1-olate)
Synonyms: titanium(4+) tetrakis(2-methylbenzenolate)
Molecular Weight (g/mol): 476.4
Molecular Formula: C28H28O4Ti
InChi Key: RLMZHOIDXMRYMJ-UHFFFAOYSA-J
SMILES: [Ti+4].CC1=CC=CC=C1[O-].CC1=CC=CC=C1[O-].CC1=CC=CC=C1[O-].CC1=CC=CC=C1[O-]
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)