accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CONNH3C

CAS RN 2847-30-5

IUPAC Name: 2-methoxy-3-methylpyrazine
Synonyms: 2-methoxy-3-methylpyrazine
Molecular Weight (g/mol): 124.14
Molecular Formula: C6H8N2O
InChi Key: VKJIAEQRKBQLLA-UHFFFAOYSA-N
SMILES: COC1=NC=CN=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results