accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 28439-86-3

IUPAC Name: 1-butoxy-4-isocyanatobenzene
Synonyms: 1-butoxy-4-isocyanatobenzene
Molecular Weight (g/mol): 191.23
Molecular Formula: C11H13NO2
InChi Key: ASFIDVOMDQBCNP-UHFFFAOYSA-N
SMILES: CCCCOC1=CC=C(C=C1)N=C=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results