CAS RN 283-60-3
CAS RN 283-60-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Silatrane, TRC
CAS: 283-60-3 Molecular Formula: C6 H13 N O3 Si Molecular Weight (g/mol): 175.26 Synonym: 2,8,9-Trioxa-5-aza-1-silabicyclo[3.3.3]undecane,NSC 626544 IUPAC Name: 4,6,11-trioxa-1-aza-5-silabicyclo[3.3.3]undecane SMILES: C1CN2CCO[SiH](OCC2)O1