accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 28249-49-2

BNOOCH3NOOHOOH

CAS RN 28249-49-2

IUPAC Name: (4-methyl-3,5-dinitrophenyl)boronic acid
Synonyms: 4-methyl-3,5-dinitrophenylboronic acid
Molecular Weight (g/mol): 225.95
Molecular Formula: C7H7BN2O6
InChi Key: DTWIWVTWNMEZOL-UHFFFAOYSA-N
SMILES: CC1=C(C=C(C=C1[N+]([O-])=O)B(O)O)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results