accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 28240-68-8

PClClPClCl

CAS RN 28240-68-8

IUPAC Name: dichloro[(dichlorophosphanyl)methyl]phosphane
Synonyms: dichloro[(dichlorophosphanyl)methyl]phosphane
Molecular Weight (g/mol): 217.77
Molecular Formula: CH2Cl4P2
InChi Key: OEKLCAXLANZKPX-UHFFFAOYSA-N
SMILES: ClP(Cl)CP(Cl)Cl
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)